ContactUsLinkComponent
Soil therm., similar DIN 58655, -15+40:0,2°C, built-in length 160mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188008-CB
Supplier:  Amarell
Soil therm., similar DIN 58655, -15+40:0,2°C, built-in length 160mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188008-CB
Supplier:  Amarell
Frequently Bought Together