ContactUsLinkComponent
Soil therm., similar DIN 58655, -25+60:0,2°C, built-in length 30mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188002-CB
Supplier:  Amarell
Soil therm., similar DIN 58655, -25+60:0,2°C, built-in length 30mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188002-CB
Supplier:  Amarell
Frequently Bought Together