Meteorological therm. BS 692, Type Minimum, with dark glass index, Min. 3/C, -50+25:0,5C, protected type, solid stem, white backed, colourless alcohol filling, round bulb with diameter max.16,5mm, in glass sheath protection, 315-345mm long, diamete 1 * 1 items
Catalog # AMARK180928-ALC
Dodavatel: Amarell
Frequently Bought Together