ContactUsLinkComponent
Meteorological therm. BS 692, Type Minimum, with dark glass index, Min. 2/C, -35+40:0,5C, protected type, solid stem, white backed, colourless alcohol filling, round bulb with diameter max.16,5mm, in glass sheath protection, 315-345mm long, diamete 1 * 1 items
Catalog # AMARK180926-ALC
Supplier:  Amarell
Meteorological therm. BS 692, Type Minimum, with dark glass index, Min. 2/C, -35+40:0,5C, protected type, solid stem, white backed, colourless alcohol filling, round bulb with diameter max.16,5mm, in glass sheath protection, 315-345mm long, diamete 1 * 1 items
Catalog # AMARK180926-ALC
Supplier:  Amarell
Frequently Bought Together