ContactUsLinkComponent
Sling psychrometer, simple, plastic, model designed for pull out, complete with 2 therm. wet-dry, -5+50:0,5C, solid stem, lens fron t, white backed, red filling, moistening tube, 20cm spare wick, evaluation table on the instrument itself 1 * 1 items
Catalog # AMARK175290-FL
Supplier:  Amarell
Sling psychrometer, simple, plastic, model designed for pull out, complete with 2 therm. wet-dry, -5+50:0,5C, solid stem, lens fron t, white backed, red filling, moistening tube, 20cm spare wick, evaluation table on the instrument itself 1 * 1 items
Catalog # AMARK175290-FL
Supplier:  Amarell
Frequently Bought Together