ContactUsLinkComponent
Sling psychrometer, solid, metal case, hinged handle, complete with 2 therm. wet-dry, -30+40:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, moistening tube, 40cm spare wick, graphical evaluation table, null
Catalog # AMARK175240-CB
Supplier:  Amarell
Sling psychrometer, solid, metal case, hinged handle, complete with 2 therm. wet-dry, -30+40:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, moistening tube, 40cm spare wick, graphical evaluation table, null
Catalog # AMARK175240-CB
Supplier:  Amarell
Frequently Bought Together