ContactUsLinkComponent
Refill therm., according to Assmann, similar DIN 58661, -35+40:0,2°C, milk glass scale, capillary tube specially coated inside, wit h non-wetting blue special liquid, total immersion, 280x8mm, without metal cap and metal ferrules, null
Catalog # AMARK175100-CB
Supplier:  Amarell
Refill therm., according to Assmann, similar DIN 58661, -35+40:0,2°C, milk glass scale, capillary tube specially coated inside, wit h non-wetting blue special liquid, total immersion, 280x8mm, without metal cap and metal ferrules, null
Catalog # AMARK175100-CB
Supplier:  Amarell
Frequently Bought Together