ContactUsLinkComponent
Meteorological Extreme therm., minimum type, blue glass index, DIN 58653, -40+40:0,5C, with glass saddle and metal cap, milk glass scale, cap. round, colourless, 290mm long, diameter 18+/-0,5mm, government verified, with Verification Certificate wit 1 * 1 items
Catalog # AMARK173040-ALC/VC
Supplier:  Amarell
Meteorological Extreme therm., minimum type, blue glass index, DIN 58653, -40+40:0,5C, with glass saddle and metal cap, milk glass scale, cap. round, colourless, 290mm long, diameter 18+/-0,5mm, government verified, with Verification Certificate wit 1 * 1 items
Catalog # AMARK173040-ALC/VC
Supplier:  Amarell
Frequently Bought Together