Precision therm., low temp., similar DIN, -38+50:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 550x6-7mm, with ring at top, null
Catalog # AMARL33104-CB
Supplier: Amarell
Frequently Bought Together