ContactUsLinkComponent
Precision therm., low temp., similar DIN, -100+30:0,2C, enclosed scale, cap. lens front, colourless, total immersion, toluene fille d, 450x9-10mm, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARL33062-TOL
Supplier:  Amarell
Precision therm., low temp., similar DIN, -100+30:0,2C, enclosed scale, cap. lens front, colourless, total immersion, toluene fille d, 450x9-10mm, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARL33062-TOL
Supplier:  Amarell
Frequently Bought Together