ContactUsLinkComponent
Precision therm., low temp., similar DIN, -58+30:0,5°C, enclosed scale, cap. lens front, colourless, total immersion, toluene filled , 300x8-9mm, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL33034-TOL
Supplier:  Amarell
Precision therm., low temp., similar DIN, -58+30:0,5°C, enclosed scale, cap. lens front, colourless, total immersion, toluene filled , 300x8-9mm, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL33034-TOL
Supplier:  Amarell
Frequently Bought Together