ContactUsLinkComponent
Precision therm., low temp., similar DIN, -150+30:1C, enclosed scale, cap. lens front, blue reflecting, total immersion, pentane co mpound, 300x8-9mm, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL33016-PENT
Supplier:  Amarell
Precision therm., low temp., similar DIN, -150+30:1C, enclosed scale, cap. lens front, blue reflecting, total immersion, pentane co mpound, 300x8-9mm, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL33016-PENT
Supplier:  Amarell
Frequently Bought Together