ContactUsLinkComponent
Refill-Therm. AMA-Exact, -10+360:1°C, enclosed scale, cap. colourless, eco-friendly, green liquid, biodegradable, richter-top finis h without button, lower part 300x8-8,5mm, upper part 350xc.10mm, suitable for government calibration, null
Catalog # AMARD265560-EF
Supplier:  Amarell
Refill-Therm. AMA-Exact, -10+360:1°C, enclosed scale, cap. colourless, eco-friendly, green liquid, biodegradable, richter-top finis h without button, lower part 300x8-8,5mm, upper part 350xc.10mm, suitable for government calibration, null
Catalog # AMARD265560-EF
Supplier:  Amarell
Frequently Bought Together