ContactUsLinkComponent
Refill-Therm. AMA-Exact, -10+150:0,5°C, enclosed scale, cap. colourless, red special liquid, richter-top finish without button, low er part 300x8-8,5mm, upper part 300xc.10mm, suitable for government calibration, null
  AMARD265552-FL
 :  Amarell
Refill-Therm. AMA-Exact, -10+150:0,5°C, enclosed scale, cap. colourless, red special liquid, richter-top finish without button, low er part 300x8-8,5mm, upper part 300xc.10mm, suitable for government calibration, null
  AMARD265552-FL
 :  Amarell
Frequently Bought Together