ContactUsLinkComponent
Refill-Therm. AMA-Exact, -10+360:1C, enclosed scale, cap. colourless, green special liquid, richter-top finish without button, low er part 250x8-8,5mm, upper part 350xc.10mm, suitable for government calibration, null
Catalog # AMARD265536-EF
Supplier:  Amarell
Refill-Therm. AMA-Exact, -10+360:1C, enclosed scale, cap. colourless, green special liquid, richter-top finish without button, low er part 250x8-8,5mm, upper part 350xc.10mm, suitable for government calibration, null
Catalog # AMARD265536-EF
Supplier:  Amarell
Frequently Bought Together