ContactUsLinkComponent
Refill-Therm. AMA-Exact, -10+250:1C, enclosed scale, cap. colourless, blue special liquid, richter-top finish without button, lowe r part 200x8-8,5mm, upper part 300xc.10mm, suitable for government calibration, null
Catalog # AMARD265508-FL
Supplier:  Amarell
Refill-Therm. AMA-Exact, -10+250:1C, enclosed scale, cap. colourless, blue special liquid, richter-top finish without button, lowe r part 200x8-8,5mm, upper part 300xc.10mm, suitable for government calibration, null
Catalog # AMARD265508-FL
Supplier:  Amarell
Frequently Bought Together