ContactUsLinkComponent
Refill therm. AMA-Exact, -10+150:1C, enclosed scale, cap. colourless, red special liquid, richter-top finish without button, lower part 200x8-8,5mm, upper part 250xc.10mm, suitable for government verification, null
Catalog # AMARD265500-FL
Supplier:  Amarell
Refill therm. AMA-Exact, -10+150:1C, enclosed scale, cap. colourless, red special liquid, richter-top finish without button, lower part 200x8-8,5mm, upper part 250xc.10mm, suitable for government verification, null
Catalog # AMARD265500-FL
Supplier:  Amarell
Frequently Bought Together