ContactUsLinkComponent
Adjustable therm. with ground joint, AMA-Exact, -10+360:1°C, enclosed scale, cap. colourless, eco-friendly, green liquid, biodegrada ble, richter-top finish without button, lower part 300x8-8,5mm, upper part 350xc.10mm, with ground joint 14,5/23 and 1 * 1 items
  AMARD265110-EF
 :  Amarell
Adjustable therm. with ground joint, AMA-Exact, -10+360:1°C, enclosed scale, cap. colourless, eco-friendly, green liquid, biodegrada ble, richter-top finish without button, lower part 300x8-8,5mm, upper part 350xc.10mm, with ground joint 14,5/23 and 1 * 1 items
  AMARD265110-EF
 :  Amarell
Frequently Bought Together