ContactUsLinkComponent
Adjustable therm. with ground joint, AMA-Exact, -10+150:0,5°C, enclosed scale, cap. colourless, red special liquid, richter-top fini sh without button, lower part 300x8-8,5mm, upper part 300xc.10mm, with ground joint 14,5/23 and universal lock, suita 1 * 1 items
Catalog # AMARD265102-FL
Supplier:  Amarell
Adjustable therm. with ground joint, AMA-Exact, -10+150:0,5°C, enclosed scale, cap. colourless, red special liquid, richter-top fini sh without button, lower part 300x8-8,5mm, upper part 300xc.10mm, with ground joint 14,5/23 and universal lock, suita 1 * 1 items
Catalog # AMARD265102-FL
Supplier:  Amarell
Frequently Bought Together