ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 73 C. white backed, -41,4-38,6:0,05C, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, with ice point scale, null
Catalog # AMARA301034-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 73 C. white backed, -41,4-38,6:0,05C, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, with ice point scale, null
Catalog # AMARA301034-CB
Supplier:  Amarell
Frequently Bought Together