ContactUsLinkComponent
Glues tester similar to Suhr, 0-56:1%mas, with thermometer, +30+85:1°C with red or blue special liquid, reference temperature 75°C, 415mm long 1 * 1 items
Catalog # AMARH860620-FL
Supplier:  Amarell
Glues tester similar to Suhr, 0-56:1%mas, with thermometer, +30+85:1°C with red or blue special liquid, reference temperature 75°C, 415mm long 1 * 1 items
Catalog # AMARH860620-FL
Supplier:  Amarell
Frequently Bought Together