ContactUsLinkComponent
Alcoholometer for destillation receiver, 0-100%vol., 330mm long, with therm. 0+35/40:1C, red or blue special liquid, divisions roun d the scale 1 * 1 items
Catalog # AMARH850298-FL
Supplier:  Amarell
Alcoholometer for destillation receiver, 0-100%vol., 330mm long, with therm. 0+35/40:1C, red or blue special liquid, divisions roun d the scale 1 * 1 items
Catalog # AMARH850298-FL
Supplier:  Amarell
Frequently Bought Together