ContactUsLinkComponent
Alcoholometer, Gay-Lussac and Cartier, 0-100/10-45, ref. temp. 15C, 280mm long, with therm. c. 0+40:1C, red or blue special liquid 1 * 1 items
Catalog # AMARH850278-FL
Supplier:  Amarell
Alcoholometer, Gay-Lussac and Cartier, 0-100/10-45, ref. temp. 15C, 280mm long, with therm. c. 0+40:1C, red or blue special liquid 1 * 1 items
Catalog # AMARH850278-FL
Supplier:  Amarell
Frequently Bought Together