ContactUsLinkComponent
Alcoholometer, similar to EUROPA class III, 85-90:0,1%vol., similar DIN 12803, with therm. +5+25:0,2C with red or blue special liqu ied, 330mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH848254-FL
Supplier:  Amarell
Alcoholometer, similar to EUROPA class III, 85-90:0,1%vol., similar DIN 12803, with therm. +5+25:0,2C with red or blue special liqu ied, 330mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH848254-FL
Supplier:  Amarell
Frequently Bought Together