Alcoholometer, similar to EUROPA class III, 80-85:0,1%vol., similar DIN 12803, with therm. +5+25:0,2C with red or blue special liqu ied, 330mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH848252-FL
Supplier: Amarell
Frequently Bought Together