ContactUsLinkComponent
Adjustable therm. with ground joint, AMA-Exact, -10+150:0,5C, enclosed scale, cap. colourless, red special liquid, richter-top fini sh without button, lower part 200x8-8,5mm, upper part 300xca.10mm, with ground joint 14,5/23 and universal lock, suit 1 * 1 items
Catalog # AMARD265054-FL
Supplier:  Amarell
Adjustable therm. with ground joint, AMA-Exact, -10+150:0,5C, enclosed scale, cap. colourless, red special liquid, richter-top fini sh without button, lower part 200x8-8,5mm, upper part 300xca.10mm, with ground joint 14,5/23 and universal lock, suit 1 * 1 items
Catalog # AMARD265054-FL
Supplier:  Amarell
Frequently Bought Together