ContactUsLinkComponent
Precision therm., high temp., similiar DIN, -10+610:1°C, enclosed scale, cap. lens front,colourless, total immersion, gallium filled , 450mm long, diameter 9-10mm, durable pigment, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL29722-GA
Supplier:  Amarell
Precision therm., high temp., similiar DIN, -10+610:1°C, enclosed scale, cap. lens front,colourless, total immersion, gallium filled , 450mm long, diameter 9-10mm, durable pigment, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL29722-GA
Supplier:  Amarell
Frequently Bought Together