ContactUsLinkComponent
Precision therm., high temp., similiar DIN, -10+480:1°C, enclosed scale, cap. colourless, total immersion, gallium filled, 450mm lon g, diameter 9-10mm, durable pigment, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL29718-GA
Supplier:  Amarell
Precision therm., high temp., similiar DIN, -10+480:1°C, enclosed scale, cap. colourless, total immersion, gallium filled, 450mm lon g, diameter 9-10mm, durable pigment, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL29718-GA
Supplier:  Amarell
Frequently Bought Together