ContactUsLinkComponent
Precision therm., high temp., similar DIN, -10+610:2°C, solid stem, frosted back, total immersion, gallium filled, 450mm long, diame ter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29608-GA
Supplier:  Amarell
Precision therm., high temp., similar DIN, -10+610:2°C, solid stem, frosted back, total immersion, gallium filled, 450mm long, diame ter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29608-GA
Supplier:  Amarell
Frequently Bought Together