ContactUsLinkComponent
Precision therm., high temp., similar DIN, -10+480:2°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diame ter, 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29006-GA
Supplier:  Amarell
Precision therm., high temp., similar DIN, -10+480:2°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diame ter, 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29006-GA
Supplier:  Amarell
Frequently Bought Together