ContactUsLinkComponent
Precision thermometer, similar DIN, -10+150:0,5C, solid stem, white backed, capillary tube specially coated inside, with non-wetti ng blue special liquid, 350x6-7mm, durable pigment, null
Catalog # AMARL27336-CB
Supplier:  Amarell
Precision thermometer, similar DIN, -10+150:0,5C, solid stem, white backed, capillary tube specially coated inside, with non-wetti ng blue special liquid, 350x6-7mm, durable pigment, null
Catalog # AMARL27336-CB
Supplier:  Amarell
Frequently Bought Together