ContactUsLinkComponent
Therm. for distillation range, similar to Kraemer-Spilker, similar to DIN 12785, enclosed scale, +75+115:0,05°C, length 500mm max., diameter 7mm, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 90mm, richter-to 1 * 1 items
Catalog # AMARD259502-CB
Supplier:  Amarell
Therm. for distillation range, similar to Kraemer-Spilker, similar to DIN 12785, enclosed scale, +75+115:0,05°C, length 500mm max., diameter 7mm, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 90mm, richter-to 1 * 1 items
Catalog # AMARD259502-CB
Supplier:  Amarell
Frequently Bought Together