Thermometer similar to AFNOR, -20+420:1°C, solid stem, white backed, gallium filled, 405mm long, total immersion, suitable for gover nment verification 1 * 1 items
Thermometer similar to AFNOR, -20+420:1°C, solid stem, white backed, gallium filled, 405mm long, total immersion, suitable for gover nment verification 1 * 1 items