ContactUsLinkComponent
Thermometer similar to AFNOR, -48+4:0,2°C, solid stem, white backed, toluene filled, 302mm long, 35mm immersion, suitable for govern ment calibration 1 * 1 items
Catalog # AMARA411480-TOL
Supplier:  Amarell
Thermometer similar to AFNOR, -48+4:0,2°C, solid stem, white backed, toluene filled, 302mm long, 35mm immersion, suitable for govern ment calibration 1 * 1 items
Catalog # AMARA411480-TOL
Supplier:  Amarell
Frequently Bought Together