ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,2/75/125, +75+125:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 375mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA411040-CB
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,2/75/125, +75+125:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 375mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA411040-CB
Supplier:  Amarell
Frequently Bought Together