ContactUsLinkComponent
Thermometer similar to BS 593, F150C/76, -20+150:1C, solid stem, white backed, capillary tube specially coated inside, with non-we tting blue special liquid, 322mm long, 76mm immersion, durable pigment, null
Catalog # AMARA403404-CB
Supplier:  Amarell
Thermometer similar to BS 593, F150C/76, -20+150:1C, solid stem, white backed, capillary tube specially coated inside, with non-we tting blue special liquid, 322mm long, 76mm immersion, durable pigment, null
Catalog # AMARA403404-CB
Supplier:  Amarell
Frequently Bought Together