ContactUsLinkComponent
Thermometer similar to BS 593, F255C/100, 95+255:0,5°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, durab le pigment, suitable for government verification 1 * 1 items
  AMARA403402-GA
 :  Amarell
Thermometer similar to BS 593, F255C/100, 95+255:0,5°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, durab le pigment, suitable for government verification 1 * 1 items
  AMARA403402-GA
 :  Amarell
Frequently Bought Together