ContactUsLinkComponent
Thermometer similar to BS 593, F300C/100, 248+302:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
  AMARA403400-GA
 :  Amarell
Thermometer similar to BS 593, F300C/100, 248+302:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
  AMARA403400-GA
 :  Amarell
Frequently Bought Together