ContactUsLinkComponent
Thermometer similar to BS 593, F225C/100, 173+227:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA403394-GA
Supplier:  Amarell
Thermometer similar to BS 593, F225C/100, 173+227:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA403394-GA
Supplier:  Amarell
Frequently Bought Together