ContactUsLinkComponent
Thermometer similar to BS 593, F150C/100, +98+152:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403388-CB
Supplier:  Amarell
Thermometer similar to BS 593, F150C/100, +98+152:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403388-CB
Supplier:  Amarell
Frequently Bought Together