Thermometer similar to BS 593, F75C/100, 24+78:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non-we tting blue special liquid, 395mm long, 100mm immersion, durable pigment null
Catalog # AMARA403382-CB
Supplier: Amarell
Frequently Bought Together