ContactUsLinkComponent
Thermometer similar to BS 593, F50C/100, -2+52:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non-we tting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403380-CB
Supplier:  Amarell
Thermometer similar to BS 593, F50C/100, -2+52:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non-we tting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403380-CB
Supplier:  Amarell
Frequently Bought Together