ContactUsLinkComponent
Thermometer similar to BS 593, B110C/100, 50+110:0,2C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400mm long, 100mm immersion, durable pigment, with ice point scale, null
Catalog # AMARA403256-CB
Supplier:  Amarell
Thermometer similar to BS 593, B110C/100, 50+110:0,2C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400mm long, 100mm immersion, durable pigment, with ice point scale, null
Catalog # AMARA403256-CB
Supplier:  Amarell
Frequently Bought Together