ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 48 F, white backed, 177,5+182,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300680-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 48 F, white backed, 177,5+182,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300680-CB
Supplier:  Amarell
Frequently Bought Together