ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 45 F, white backed, 74,5+79,5:0,1F, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300650-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 45 F, white backed, 74,5+79,5:0,1F, capillary tube specially coated inside, with non-wett ing blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300650-CB
Supplier:  Amarell
Frequently Bought Together