ContactUsLinkComponent
Thermometer similar to ASTM 43 C, solid stem, white backed, -51,6-34:0,1°C, toluene filled, total immersion, 420x7-8mm, suitable for government calibration, with ice point scale 1 * 1 items
Catalog # AMARA300628-TOL
Supplier:  Amarell
Thermometer similar to ASTM 43 C, solid stem, white backed, -51,6-34:0,1°C, toluene filled, total immersion, 420x7-8mm, suitable for government calibration, with ice point scale 1 * 1 items
Catalog # AMARA300628-TOL
Supplier:  Amarell
Frequently Bought Together