ContactUsLinkComponent
Clarification hydrometer, 0-18:0,5%, without therm., reading above, ref. temp. 70C, 290mm long 1 * 1 items
Catalog # AMARH847494
Supplier:  Amarell
Clarification hydrometer, 0-18:0,5%, without therm., reading above, ref. temp. 70C, 290mm long 1 * 1 items
Catalog # AMARH847494
Supplier:  Amarell
Frequently Bought Together