ContactUsLinkComponent
Thermometer similar to BS 593, A100C/100, 70+100:0,1C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400mm long, 100mm immersion, durable pigment, with ice point scale, null
Catalog # AMARA403248-CB
Supplier:  Amarell
Thermometer similar to BS 593, A100C/100, 70+100:0,1C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400mm long, 100mm immersion, durable pigment, with ice point scale, null
Catalog # AMARA403248-CB
Supplier:  Amarell
Frequently Bought Together