ContactUsLinkComponent
Thermometer similar to BS 593, A20C/100, -20+20:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403242-CB
Supplier:  Amarell
Thermometer similar to BS 593, A20C/100, -20+20:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403242-CB
Supplier:  Amarell
Frequently Bought Together