ContactUsLinkComponent
Thermometer similar to BS 593, A10C/100, -30+10:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Katalogové číslo AMARA403240-CB
Dodavatel:  Amarell
Thermometer similar to BS 593, A10C/100, -30+10:0,1C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Katalogové číslo AMARA403240-CB
Dodavatel:  Amarell
Frequently Bought Together